Product Details

SNP ID
hCV175592421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49031382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCCTCTTCCCCCACCACCAT[A/C]TCCTCTTTTCAAACCTGAATTCGGA
Phenotype
MIM: 314310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TFE3 PubMed Links

Gene Details

Gene
TFE3
Gene Name
transcription factor binding to IGHM enhancer 3
There are no transcripts associated with this gene.

View Full Product Details