Product Details

SNP ID
rs5257
Assay Type
Functionally tested
NCBI dbSNP Submissions
86
Location
Chr.1:16046629 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGGCCCTCGTCTCTTTCTCTTC[A/G]GGCTTCTCTCAGAGCATCACACCCT
Phenotype
MIM: 602023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 476 Silent Mutation TCA,TCG S108S NP_000076.2
NM_001165945.2 476 Intron NP_001159417.2

View Full Product Details