Product Details

SNP ID
rs11539636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50362963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACAAAATAGAACAACAAAAAAA[A/G]GGCAGAGAAAAGGAAATGGCCCCCC
Phenotype
MIM: 607082 MIM: 607068 MIM: 607069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA2D2 PubMed Links

Gene Details

Gene
CACNA2D2
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005505.2 5111 UTR 3 NP_001005505.1
NM_001174051.2 5111 UTR 3 NP_001167522.1
NM_001291101.1 5111 UTR 3 NP_001278030.1
NM_006030.3 5111 UTR 3 NP_006021.2
XM_005265581.4 5111 UTR 3 XP_005265638.1
XM_011534243.2 5111 UTR 3 XP_011532545.1
Gene
CYB561D2
Gene Name
cytochrome b561 family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291284.1 5111 Intron NP_001278213.1
NM_007022.4 5111 Intron NP_008953.1
Gene
TMEM115
Gene Name
transmembrane protein 115
There are no transcripts associated with this gene.

View Full Product Details