Product Details

SNP ID
hCV176986316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102515140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACCTGTCAAACTTCTCCACTTC[C/T]GACAAGTCTGGCTTATCACTCATCT
Phenotype
MIM: 300939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMSB15A PubMed Links

Gene Details

Gene
TMSB15A
Gene Name
thymosin beta 15a
There are no transcripts associated with this gene.

View Full Product Details