Product Details

SNP ID
rs184177701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102341604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAACACTCTTCAGACCAGGAC[A/G]CCTGGGACAGCCTGAGGAGCCCGGG
Phenotype
MIM: 613362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CINP PubMed Links

Gene Details

Gene
CINP
Gene Name
cyclin dependent kinase 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320046.1 2074 Intron NP_001306975.1
NM_032630.2 2074 Intron NP_116019.1
Gene
ZNF839
Gene Name
zinc finger protein 839
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267827.1 2074 Missense Mutation ACC,GCC T621A NP_001254756.1
NM_001267828.1 2074 Missense Mutation ACC,GCC T621A NP_001254757.1
NM_018335.4 2074 Missense Mutation ACC,GCC T737A NP_060805.3
XM_006720203.3 2074 Missense Mutation ACC,GCC T649A XP_006720266.2
XM_011536945.1 2074 Missense Mutation ACC,GCC T771A XP_011535247.1
XM_011536946.2 2074 Missense Mutation ACC,GCC T721A XP_011535248.1
XM_011536948.2 2074 Missense Mutation ACC,GCC T621A XP_011535250.1
XM_011536949.2 2074 Intron XP_011535251.1
XM_011536950.2 2074 Intron XP_011535252.1
XM_017021447.1 2074 Missense Mutation ACC,GCC T687A XP_016876936.1
XM_017021448.1 2074 Intron XP_016876937.1
XM_017021449.1 2074 Missense Mutation ACC,GCC T402A XP_016876938.1
XM_017021450.1 2074 Intron XP_016876939.1
XM_017021451.1 2074 Intron XP_016876940.1

View Full Product Details