Product Details
- SNP ID
-
rs184025907
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:72074335 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCAAATGGCTATGTGGAGCACT[C/T]GTTTCGCTACCAGTGTAAGAACTAC
- Phenotype
-
MIM: 140210
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HPR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7202157,rs7202158] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HPR
- Gene Name
- haptoglobin-related protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020995.3 |
173 |
Missense Mutation |
TCG,TTG |
S48L |
NP_066275.3 |
- Gene
- TXNL4B
- Gene Name
- thioredoxin like 4B
View Full Product Details