Product Details
- SNP ID
-
rs184039815
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:32097152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTGTACTGCAGGAAGATTG[C/T]TGATCGAATCGACAATGATGGGGAT
- Phenotype
-
MIM: 602735
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RCN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs564799768] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RCN1
- Gene Name
- reticulocalbin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002901.2 |
529 |
Missense Mutation |
GCT,GTT |
A88V |
NP_002892.1 |
View Full Product Details