Product Details

SNP ID
rs185917906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31076339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTTGCCCCATGGAGGACACACC[C/T]CCCTCACTCAGCTGCTCCGACTGTC
Phenotype
MIM: 610561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 1083 Silent Mutation CCC,CCT P5P NP_055514.3
XM_005255710.3 1083 Silent Mutation CCC,CCT P5P XP_005255767.1
XM_005255711.3 1083 Silent Mutation CCC,CCT P5P XP_005255768.1
XM_005255712.3 1083 Silent Mutation CCC,CCT P5P XP_005255769.1
XM_011545990.2 1083 Silent Mutation CCC,CCT P5P XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
There are no transcripts associated with this gene.

View Full Product Details