Product Details

SNP ID
rs186169944
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104887274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAATTTGGGAAATGAAGACAGA[A/G]CCATTTGTTTCATGTCTCCAAGAAA
Phenotype
MIM: 608633
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CASP12 PubMed Links
Additional Information
For this assay, SNP(s) [rs142269030] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CASP12
Gene Name
caspase 12 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191016.2 1105 Silent Mutation GGC,GGT G283G NP_001177945.2

View Full Product Details