Product Details

SNP ID
rs187109973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:140783698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGTCTTCCAGAAAAATCTACG[C/T]CTTCCACCAAATCCAGGTCTTCCAG
Phenotype
MIM: 302650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDR1 PubMed Links

Gene Details

Gene
CDR1
Gene Name
cerebellar degeneration related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004065.2 861 Silent Mutation AGA,AGG R223R NP_004056.2

View Full Product Details