Product Details
- SNP ID
-
rs187019439
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:88133602 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGCAGAGCCTTACCTCTCAGA[C/T]TGTGGTTTTTCATCCTGCCTCTGGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107987092
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202054946,rs531505563] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107987092
- Gene Name
- spermatogenesis-associated protein 31C2-like
There are no transcripts associated with this gene.
- Gene
- SPATA31C2
- Gene Name
- SPATA31 subfamily C member 2
View Full Product Details