Product Details

SNP ID
rs189903941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111876024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGAACACTTCAGCCTTTTTAGC[A/G]GGAAAACTTTGCCTAATTCAGCAAT
Phenotype
MIM: 606941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG9 PubMed Links

Gene Details

Gene
ALG9
Gene Name
ALG9, alpha-1,2-mannosyltransferase
There are no transcripts associated with this gene.

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
There are no transcripts associated with this gene.

Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138378.2 1098 Missense Mutation CCG,CTG P258L NP_612387.1

View Full Product Details