Product Details

SNP ID
rs189486852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:126164959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGACGTGGGACTGAGAGCCCAC[A/G]GCGTACAGGGACAACTCATCACAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF12L2 PubMed Links

Gene Details

Gene
DCAF12L2
Gene Name
DDB1 and CUL4 associated factor 12 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013628.2 1139 Silent Mutation GCC,GCT A322A NP_001013650.1

View Full Product Details