Product Details

SNP ID
rs190100022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56359011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCGCCTAGCTGCGCACCATGGC[C/T]GGGTTGCTGGCGTTGCTGGGTCCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM210B PubMed Links

Gene Details

Gene
FAM210B
Gene Name
family with sequence similarity 210 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080821.2 85 Silent Mutation GCC,GCT A2A NP_543011.2

View Full Product Details