Product Details
- SNP ID
-
rs190564988
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39451977 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGGACTGCCGGAGCCCAGTGGAC[C/T]CCTGGGCTGCTGCCAGCCCCGACCT
- Phenotype
-
MIM: 602230
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LCA5L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11575939] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCA5L
- Gene Name
- LCA5L, lebercilin like
There are no transcripts associated with this gene.
- Gene
- MIR6508
- Gene Name
- microRNA 6508
There are no transcripts associated with this gene.
- Gene
- SH3BGR
- Gene Name
- SH3 domain binding glutamate rich protein
- Gene
- WRB-SH3BGR
- Gene Name
- WRB-SH3BGR readthrough
View Full Product Details