Product Details

SNP ID
rs191515075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18465684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTAGTGATTATCACAGCAGGT[A/G]CACGCCAGAAAAAAGGAGAAACACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LDHAL6A PubMed Links

Gene Details

Gene
LDHAL6A
Gene Name
lactate dehydrogenase A like 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144071.1 381 Missense Mutation ACA,GCA T98A NP_001137543.1
NM_144972.4 381 Missense Mutation ACA,GCA T98A NP_659409.2
XM_005252805.4 381 Missense Mutation ACA,GCA T122A XP_005252862.2
XM_011519922.2 381 Missense Mutation ACA,GCA T122A XP_011518224.1
XM_011519923.2 381 Missense Mutation ACA,GCA T98A XP_011518225.1
XM_011519924.2 381 Missense Mutation ACA,GCA T98A XP_011518226.1
XM_011519925.1 381 Missense Mutation ACA,GCA T98A XP_011518227.1

View Full Product Details