Product Details

SNP ID
rs191303228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6240330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGATGCTTTACATTTTTGTCG[G/T]CATCCATTGGAACAGCTGCCTATAC
Phenotype
MIM: 609472
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNGA4 PubMed Links

Gene Details

Gene
CNGA4
Gene Name
cyclic nucleotide gated channel alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037329.3 558 Missense Mutation GGC,GTC G179V NP_001032406.1
XM_017017217.1 558 Missense Mutation GGC,GTC G179V XP_016872706.1
XM_017017218.1 558 Missense Mutation GGC,GTC G139V XP_016872707.1
XM_017017219.1 558 Missense Mutation GGC,GTC G139V XP_016872708.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
There are no transcripts associated with this gene.

View Full Product Details