Product Details

SNP ID
rs7159215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92922775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGACCGGGGGAGCATACCCCAG[A/G]CAGAGGGCACTTAGGAAAAATAGCA
Phenotype
MIM: 118910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001275.3 187 Intron NP_001266.1
NM_001301690.1 187 Intron NP_001288619.1
XM_011536370.1 187 UTR 5 XP_011534672.1

View Full Product Details