Product Details

SNP ID
rs7443900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:175660108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGATGCGTCCGTGGGAAAAACAA[A/G]ACCAGCTGTGGTCCATCTCTGCCAA
Phenotype
MIM: 142703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRH2 PubMed Links

Gene Details

Gene
HRH2
Gene Name
histamine receptor H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131055.1 496 Intron NP_001124527.1
NM_022304.2 496 Intron NP_071640.1
XM_005265904.3 496 Intron XP_005265961.1
XM_006714864.3 496 UTR 5 XP_006714927.1
XM_006714865.3 496 Intron XP_006714928.1
XM_011534548.2 496 UTR 5 XP_011532850.1
XM_011534549.2 496 UTR 5 XP_011532851.1
XM_017009426.1 496 Intron XP_016864915.1

View Full Product Details