Product Details

SNP ID
rs7553830
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:193066727 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATATATAAACATGTTCAAGTAA[A/G]CATGATCTTCAGAACAAAACTTCTT
Phenotype
MIM: 600063 MIM: 610667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCARNA18B PubMed Links

Gene Details

Gene
SCARNA18B
Gene Name
small Cajal body-specific RNA 18B
There are no transcripts associated with this gene.

Gene
TROVE2
Gene Name
TROVE domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042369.2 Intron NP_001035828.1
NM_001042370.2 Intron NP_001035829.2
NM_001173524.1 Intron NP_001166995.1
NM_001173525.1 Intron NP_001166996.1
NM_004600.5 Intron NP_004591.2
XM_006711495.3 Intron XP_006711558.1
XM_006711496.3 Intron XP_006711559.1
XM_006711497.3 Intron XP_006711560.1
XM_011509922.2 Intron XP_011508224.1
XM_017002180.1 Intron XP_016857669.1
XM_017002181.1 Intron XP_016857670.1
XM_017002182.1 Intron XP_016857671.1
XM_017002183.1 Intron XP_016857672.1
Gene
UCHL5
Gene Name
ubiquitin C-terminal hydrolase L5
There are no transcripts associated with this gene.

View Full Product Details