Product Details
- SNP ID
-
rs968402
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:233868105 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCTGGCCAATCTACGATTTTCACT[G/A]GGCTCACGAAGCACATGATCTTCGG
- Phenotype
-
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MSL3P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13390675] are located under a probe and SNP(s) [rs16849914] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSL3P1
- Gene Name
- male-specific lethal 3 homolog (Drosophila) pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details