Product Details

SNP ID
rs201710528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133309780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCACGTCTCCTTGGCAGGGTG[C/G]CCGCGTTTGCCTGCCAGGTGGCCGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TUBGCP2 PubMed Links

Gene Details

Gene
TUBGCP2
Gene Name
tubulin gamma complex associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256617.1 287 Intron NP_001243546.1
NM_001256618.1 287 Intron NP_001243547.1
NM_006659.3 287 Intron NP_006650.1
XM_017015595.1 287 Intron XP_016871084.1
XM_017015596.1 287 Intron XP_016871085.1
Gene
ZNF511
Gene Name
zinc finger protein 511
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145806.3 287 Missense Mutation CCC,GCC P78A NP_665805.2

View Full Product Details