Product Details

SNP ID
rs200119903
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6449399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATTGAACTGCCCATTGCCCTC[A/G]CCATGGGAGCCAAACTTGAAGAGGA
Phenotype
MIM: 142290 MIM: 605493
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HPX PubMed Links

Gene Details

Gene
HPX
Gene Name
hemopexin
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 2165 Silent Mutation GGC,GGT G663G NP_001234935.1
NM_001248007.1 2165 Silent Mutation GGC,GGT G544G NP_001234936.1
NM_006458.3 2165 Silent Mutation GGC,GGT G663G NP_006449.2
NM_033278.3 2165 Silent Mutation GGC,GGT G663G NP_150594.2
XM_011519844.1 2165 Silent Mutation GGC,GGT G663G XP_011518146.1
XM_011519845.2 2165 Silent Mutation GGC,GGT G544G XP_011518147.1
XM_017017098.1 2165 Silent Mutation GGC,GGT G663G XP_016872587.1
XM_017017099.1 2165 Silent Mutation GGC,GGT G544G XP_016872588.1

View Full Product Details