Product Details

SNP ID
rs201667418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64231902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGTACTTATTATGAACTGTTG[G/T]GGGTGCATCCTGGTGCCAGCACTGA
Phenotype
MIM: 604189 MIM: 607901 MIM: 186946 MIM: 610470 MIM: 601398
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307980.1 541 Missense Mutation GGG,TGG G40W NP_001294909.1
NM_001307981.1 541 Missense Mutation GGG,TGG G40W NP_001294910.1
NM_005528.3 541 Missense Mutation GGG,TGG G40W NP_005519.2
Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
FKBP2
Gene Name
FK506 binding protein 2
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
There are no transcripts associated with this gene.

Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
There are no transcripts associated with this gene.

Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details