Product Details

SNP ID
rs202145026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65961810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGTCCAAGAAGCATCGCGGAGA[C/G]AAGGAGGCGGCCGGGACGACGGCGG
Phenotype
MIM: 605941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SART1 PubMed Links

Gene Details

Gene
SART1
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005146.4 83 Missense Mutation GAC,GAG D10E NP_005137.1
XM_011545344.1 83 UTR 5 XP_011543646.1
XM_011545345.2 83 Intron XP_011543647.1
Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
There are no transcripts associated with this gene.

View Full Product Details