Product Details

SNP ID
rs199646358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124326195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCCCACCTGGTGCCCACCTGC[A/G]GACGAGGGCCTGTCCTCCCACACGC
Phenotype
MIM: 600848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCOR2 PubMed Links

Gene Details

Gene
NCOR2
Gene Name
nuclear receptor corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077261.3 7512 Silent Mutation TCC,TCT S2397S NP_001070729.2
NM_001206654.1 7512 Silent Mutation TCC,TCT S2443S NP_001193583.1
NM_006312.5 7512 Silent Mutation TCC,TCT S2453S NP_006303.4

View Full Product Details