Product Details

SNP ID
rs201197821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102342054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGAGTTACTGTCTCAGGGACAG[A/C]AGCAGATTTTTATTCAGACTTCCGA
Phenotype
MIM: 613362
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CINP PubMed Links

Gene Details

Gene
CINP
Gene Name
cyclin dependent kinase 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320046.1 2524 Intron NP_001306975.1
NM_032630.2 2524 Intron NP_116019.1
Gene
ZNF839
Gene Name
zinc finger protein 839
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267827.1 2524 Missense Mutation AAG,CAG K771Q NP_001254756.1
NM_001267828.1 2524 Missense Mutation AAG,CAG K771Q NP_001254757.1
NM_018335.4 2524 Missense Mutation AAG,CAG K887Q NP_060805.3
XM_006720203.3 2524 Missense Mutation AAG,CAG K799Q XP_006720266.2
XM_011536945.1 2524 Missense Mutation AAG,CAG K921Q XP_011535247.1
XM_011536946.2 2524 Missense Mutation AAG,CAG K871Q XP_011535248.1
XM_011536948.2 2524 Missense Mutation AAG,CAG K771Q XP_011535250.1
XM_011536949.2 2524 Intron XP_011535251.1
XM_011536950.2 2524 Intron XP_011535252.1
XM_017021447.1 2524 Missense Mutation AAG,CAG K837Q XP_016876936.1
XM_017021448.1 2524 Intron XP_016876937.1
XM_017021449.1 2524 Missense Mutation AAG,CAG K552Q XP_016876938.1
XM_017021450.1 2524 Intron XP_016876939.1
XM_017021451.1 2524 Intron XP_016876940.1

View Full Product Details