Product Details

SNP ID
rs201495311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3089998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGGAAGCTCTGGCCGCAGTGG[G/T]AGCACAGGACATCGGTCAGTGGCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 1363 Missense Mutation TAC,TCC Y85S NP_001269344.1
NM_001282416.1 1363 Missense Mutation TAC,TCC Y342S NP_001269345.1
NM_032805.2 1363 Missense Mutation TAC,TCC Y479S NP_116194.2
XM_017023791.1 1363 Missense Mutation TAC,TCC Y479S XP_016879280.1
XM_017023792.1 1363 Missense Mutation TAC,TCC Y447S XP_016879281.1

View Full Product Details