Product Details

SNP ID
rs201991078
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11254864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGTCAAATCTGGAAGGGGAAGGA[C/G]CTCAGGTAGTCGCGGAGGACGGGGT
Phenotype
MIM: 603597
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SOCS1 PubMed Links

Gene Details

Gene
SOCS1
Gene Name
suppressor of cytokine signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003745.1 769 Silent Mutation AGC,AGG S205R NP_003736.1

View Full Product Details