Product Details

SNP ID
rs201674604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67230464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCATGATGCGCACTTCACGGGC[C/T]GCCTGCGGGGTGTAGCCCAGGTAGA
Phenotype
MIM: 606881
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318202.1 3307 Silent Mutation GCA,GCG A993A NP_001305131.1
NM_013241.2 3307 Silent Mutation GCA,GCG A967A NP_037373.2
XM_006721180.1 3307 Silent Mutation GCA,GCG A498A XP_006721243.1
XM_011523043.2 3307 Silent Mutation GCA,GCG A983A XP_011521345.1
XM_011523044.1 3307 Silent Mutation GCA,GCG A939A XP_011521346.1
XM_011523045.2 3307 Silent Mutation GCA,GCG A900A XP_011521347.1
Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
TMEM208
Gene Name
transmembrane protein 208
There are no transcripts associated with this gene.

View Full Product Details