Product Details

SNP ID
rs200854554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9022993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGCCGTGTGCACCGACTCGC[A/G]CACCGACATGCGCCCGCTCTCGGGC
Phenotype
MIM: 601614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTN1 PubMed Links

Gene Details

Gene
NTN1
Gene Name
netrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004822.2 827 Missense Mutation CAC,CGC H207R NP_004813.2
XM_006721595.3 827 Missense Mutation CAC,CGC H207R XP_006721658.1

View Full Product Details