Product Details

SNP ID
rs201493311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17249015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATACCGTCCTTCTGGTTAATAC[C/T]TGCAAAAATCTCACCATCTTCTATC
Phenotype
MIM: 604665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS3 PubMed Links

Gene Details

Gene
COPS3
Gene Name
COP9 signalosome subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199125.1 1023 Missense Mutation AGT,GGT S330G NP_001186054.1
NM_001316354.1 1023 Missense Mutation AGT,GGT S220G NP_001303283.1
NM_001316355.1 1023 Missense Mutation AGT,GGT S292G NP_001303284.1
NM_001316356.1 1023 Missense Mutation AGT,GGT S283G NP_001303285.1
NM_001316357.1 1023 Missense Mutation AGT,GGT S245G NP_001303286.1
NM_001316358.1 1023 Missense Mutation AGT,GGT S245G NP_001303287.1
NM_003653.3 1023 Missense Mutation AGT,GGT S350G NP_003644.2
XM_005256837.4 1023 Missense Mutation AGT,GGT S330G XP_005256894.1
XM_005256840.3 1023 Missense Mutation AGT,GGT S220G XP_005256897.1
XM_005256842.4 1023 Missense Mutation AGT,GGT S220G XP_005256899.1
XM_017025246.1 1023 Missense Mutation AGT,GGT S245G XP_016880735.1

View Full Product Details