Product Details

SNP ID
rs199749467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50154675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCCAACAGCGCCTGGTTCCTC[A/G]GGTATTTGCTGTCCATCTGGTACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IZUMO2 PubMed Links

Gene Details

Gene
IZUMO2
Gene Name
IZUMO family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321449.1 635 Missense Mutation CCG,CTG P183L NP_001308378.1
NM_152358.2 635 Missense Mutation CCG,CTG P183L NP_689571.2
XM_011526436.2 635 Silent Mutation CCC,CCT P173P XP_011524738.1
XM_011526437.2 635 Silent Mutation CCC,CCT P166P XP_011524739.1

View Full Product Details