Product Details

SNP ID
rs200375319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45408244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGTCCCCAAGCTGGAGAAGC[A/G]ACCCTGCTGGCCCCCTCAACGGAGG
Phenotype
MIM: 107325 MIM: 126380 MIM: 607463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD3EAP PubMed Links

Gene Details

Gene
CD3EAP
Gene Name
CD3e molecule associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297590.1 2399 Silent Mutation GCA,GCG A94A NP_001284519.1
NM_012099.1 2399 Silent Mutation GCA,GCG A92A NP_036231.1
XM_017026180.1 2399 UTR 5 XP_016881669.1
XM_017026181.1 2399 UTR 5 XP_016881670.1
Gene
ERCC1
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166049.1 2399 UTR 3 NP_001159521.1
NM_001983.3 2399 UTR 3 NP_001974.1
NM_202001.2 2399 Intron NP_973730.1
XM_005258634.1 2399 Intron XP_005258691.1
XM_005258635.2 2399 Intron XP_005258692.1
XM_005258636.4 2399 Intron XP_005258693.1
XM_005258637.1 2399 Intron XP_005258694.1
XM_011526610.2 2399 Intron XP_011524912.1
XM_017026459.1 2399 Intron XP_016881948.1
XM_017026460.1 2399 UTR 3 XP_016881949.1
XM_017026461.1 2399 UTR 3 XP_016881950.1
XM_017026462.1 2399 UTR 3 XP_016881951.1
XM_017026463.1 2399 UTR 3 XP_016881952.1
XM_017026464.1 2399 UTR 3 XP_016881953.1
XM_017026465.1 2399 UTR 3 XP_016881954.1
XM_017026466.1 2399 UTR 3 XP_016881955.1
Gene
PPP1R13L
Gene Name
protein phosphatase 1 regulatory subunit 13 like
There are no transcripts associated with this gene.

View Full Product Details