Product Details

SNP ID
rs200445019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3594865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCTGGACAGAGCCTTCCCTG[C/T]TGGAGGTTCAAAAGGAAGCAACTGT
Phenotype
MIM: 608792 MIM: 188070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIPC3 PubMed Links

Gene Details

Gene
GIPC3
Gene Name
GIPC PDZ domain containing family member 3
There are no transcripts associated with this gene.

Gene
TBXA2R
Gene Name
thromboxane A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001060.5 1582 UTR 3 NP_001051.1
NM_201636.2 1582 Missense Mutation ACA,GCA T399A NP_963998.2
XM_011528214.2 1582 Intron XP_011526516.1

View Full Product Details