Product Details

SNP ID
rs201171834
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:158480067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGTCATTCTTAGTGGCAATGTC[A/G]CCATTATCAGTGTCATCCACCTGGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 190 Missense Mutation ACC,GCC T64A NP_001004472.1

View Full Product Details