Product Details

SNP ID
rs201662493
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:36341836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGTCGTGGCCCAGCCGTCGCAC[C/T]GGTGGTGCGCTGCCGAACTGCCGCT
Phenotype
MIM: 609437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK40 PubMed Links

Gene Details

Gene
STK40
Gene Name
serine/threonine kinase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282546.1 1677 Silent Mutation CCA,CCG P414P NP_001269475.1
NM_001282547.1 1677 Silent Mutation CCA,CCG P409P NP_001269476.1
NM_032017.2 1677 Silent Mutation CCA,CCG P409P NP_114406.1

View Full Product Details