Product Details

SNP ID
rs201080691
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:158480109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTGGATAAAAGCCTCCACACA[C/T]CAATGTACTTCTTCCTTGGCATTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 232 Missense Mutation CCA,TCA P78S NP_001004472.1

View Full Product Details