Product Details

SNP ID
rs200464534
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:16047918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCGCCAAGGTTCTGGAATCCC[A/G]GAGGTGAAGACCATGTTGGCGGGTG
Phenotype
MIM: 602023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 524 Silent Mutation CCA,CCG P124P NP_000076.2
NM_001165945.2 524 Intron NP_001159417.2
Gene
FAM131C
Gene Name
family with sequence similarity 131 member C
There are no transcripts associated with this gene.

View Full Product Details