Product Details

SNP ID
rs200173579
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:17372255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCAGGCCTGAGGATGGTCAGC[A/G]TGGAGGGCCGAGCCATGTCCTTCCA
Phenotype
MIM: 605347 MIM: 610363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI4 PubMed Links

Gene Details

Gene
PADI4
Gene Name
peptidyl arginine deiminase 4
There are no transcripts associated with this gene.

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207421.4 60 Missense Mutation ATG,GTG M4V NP_997304.3

View Full Product Details