Product Details

SNP ID
rs202157454
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:74706325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAGAATCATTTTTCCAGTAG[C/T]CCCACTACCATGAATGATATTTTCA
Phenotype
MIM: 123691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 950 Missense Mutation ACT,GCT T321A NP_001123514.1
NM_001130043.1 950 Missense Mutation ACT,GCT T287A NP_001123515.1
NM_001134759.1 950 Missense Mutation ACT,GCT T184A NP_001128231.1
NM_001889.3 950 Missense Mutation ACT,GCT T321A NP_001880.2
XM_005270491.4 950 Missense Mutation ACT,GCT T184A XP_005270548.1
XM_011540747.1 950 Missense Mutation ACT,GCT T321A XP_011539049.1
XM_017000367.1 950 Missense Mutation ACT,GCT T287A XP_016855856.1
XM_017000368.1 950 Missense Mutation ACT,GCT T184A XP_016855857.1

View Full Product Details