Product Details

SNP ID
rs199682548
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45534468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTATGAGCCAAATCCTGGAGGT[C/T]ATTCAAGCTCCTCCTTATGGTATAA
Phenotype
MIM: 609031
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EPPIN PubMed Links

Gene Details

Gene
EPPIN
Gene Name
epididymal peptidase inhibitor
There are no transcripts associated with this gene.

Gene
EPPIN-WFDC6
Gene Name
EPPIN-WFDC6 readthrough
There are no transcripts associated with this gene.

Gene
WFDC6
Gene Name
WAP four-disulfide core domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080827.1 348 Silent Mutation TAA,TGA *87* NP_543017.1

View Full Product Details