Product Details
- SNP ID
-
rs199990768
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:2483893 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCGCTAAAGCCTCGCCCACACTC[A/G]CTGCAAACATAGGGCTTCTCCTCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF343
PubMed Links
Gene Details
- Gene
- ZNF343
- Gene Name
- zinc finger protein 343
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282495.1 |
1420 |
Silent Mutation |
AGC,AGT |
S356S |
NP_001269424.1 |
NM_001282496.1 |
1420 |
Silent Mutation |
AGC,AGT |
S356S |
NP_001269425.1 |
NM_001282497.1 |
1420 |
Silent Mutation |
AGC,AGT |
S397S |
NP_001269426.1 |
NM_001282498.1 |
1420 |
Silent Mutation |
AGC,AGT |
S266S |
NP_001269427.1 |
NM_001282499.1 |
1420 |
Intron |
|
|
NP_001269428.1 |
NM_001321800.1 |
1420 |
Silent Mutation |
AGC,AGT |
S356S |
NP_001308729.1 |
NM_001321801.1 |
1420 |
Silent Mutation |
AGC,AGT |
S397S |
NP_001308730.1 |
NM_001321802.1 |
1420 |
Silent Mutation |
AGC,AGT |
S396S |
NP_001308731.1 |
NM_001321803.1 |
1420 |
Silent Mutation |
AGC,AGT |
S396S |
NP_001308732.1 |
NM_001321805.1 |
1420 |
Silent Mutation |
AGC,AGT |
S355S |
NP_001308734.1 |
NM_024325.5 |
1420 |
Silent Mutation |
AGC,AGT |
S356S |
NP_077301.4 |
XM_017028062.1 |
1420 |
Silent Mutation |
AGC,AGT |
S356S |
XP_016883551.1 |
View Full Product Details