Product Details
- SNP ID
-
rs201478144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38427129 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCACCTCGTAGGTCTTGTGAAAA[C/T]GTGAGTAGTCCACCTTGTAGTGGCT
- Phenotype
-
MIM: 600504
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNJ4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs196059] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNJ4
- Gene Name
- potassium voltage-gated channel subfamily J member 4
- Gene
- LOC101927183
- Gene Name
- uncharacterized LOC101927183
There are no transcripts associated with this gene.
View Full Product Details