Product Details

SNP ID
rs200438719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19433276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTATCTTTCAGGCTTCTGGGAA[C/T]TTGGCAGACGCAGCTTAGAGAGACT
Phenotype
MIM: 600237 MIM: 605089
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
There are no transcripts associated with this gene.

Gene
HIRA
Gene Name
histone cell cycle regulator
There are no transcripts associated with this gene.

Gene
MRPL40
Gene Name
mitochondrial ribosomal protein L40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318151.1 718 UTR 5 NP_001305080.1
NM_001318152.1 718 UTR 5 NP_001305081.1
NM_003776.3 718 Missense Mutation ACT,ATT T22I NP_003767.2

View Full Product Details