Product Details

SNP ID
rs200691173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48687922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTCTTTGGAATGTCTTAGTGAA[A/T]ATTGCATACAGAAATGGATTGGCAC
Phenotype
MIM: 605358 MIM: 152790
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GTF2A1L PubMed Links

Gene Details

Gene
GTF2A1L
Gene Name
general transcription factor IIA subunit 1 like
There are no transcripts associated with this gene.

Gene
LHCGR
Gene Name
luteinizing hormone/choriogonadotropin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000233.3 1671 Silent Mutation ATA,ATT I625I NP_000224.2
XM_005264309.3 1671 Silent Mutation ATA,ATT I306I XP_005264366.1
XM_006712015.3 1671 Silent Mutation ATA,ATT I315I XP_006712078.1
XM_011532828.1 1671 Silent Mutation ATA,ATT I600I XP_011531130.1
XM_011532831.1 1671 Silent Mutation ATA,ATT I413I XP_011531133.1
XM_011532834.2 1671 Silent Mutation ATA,ATT I306I XP_011531136.1
XM_017004089.1 1671 Silent Mutation ATA,ATT I540I XP_016859578.1
XM_017004090.1 1671 Silent Mutation ATA,ATT I413I XP_016859579.1
Gene
STON1-GTF2A1L
Gene Name
STON1-GTF2A1L readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198593.1 1671 Intron NP_001185522.1
NM_001198594.1 1671 Intron NP_001185523.1
NM_172311.2 1671 Intron NP_758515.1

View Full Product Details