Product Details

SNP ID
rs199571394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108378496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGACCTGCTTATTTCTACCTA[A/T]CCTAAAGCAGGTAGGTGGAAGGGCT
Phenotype
MIM: 608357
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SULT1C4 PubMed Links

Gene Details

Gene
SULT1C4
Gene Name
sulfotransferase family 1C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321770.1 586 Silent Mutation TAA,TAT *53Y NP_001308699.1
NM_006588.3 586 Silent Mutation TAA,TAT *53Y NP_006579.2
XM_017003807.1 586 UTR 5 XP_016859296.1

View Full Product Details