Product Details

SNP ID
rs202160441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240029533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCCCAAGGCCTTTTTCAAGGC[A/G]TCCTTAAATTCCTTGTTCCTCAGGC
Phenotype
MIM: 603835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA10 PubMed Links

Gene Details

Gene
NDUFA10
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
There are no transcripts associated with this gene.

Gene
OR6B2
Gene Name
olfactory receptor family 6 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005853.1 897 Silent Mutation GAC,GAT D299D NP_001005853.1

View Full Product Details