Product Details

SNP ID
rs202217452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26346211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCCGGGAGTCGCTGCCGAGTG[A/G]GCGCTCAGTTTTCGGGTCGTCATGG
Phenotype
MIM: 607915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRF3 PubMed Links

Gene Details

Gene
ADGRF3
Gene Name
adhesion G protein-coupled receptor F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145168.1 532 Intron NP_001138640.1
NM_001145169.1 532 Intron NP_001138641.1
NM_001321971.1 532 Silent Mutation GCC,GCT A8A NP_001308900.1
NM_001321975.1 532 Intron NP_001308904.1
NM_153835.3 532 Silent Mutation GCC,GCT A8A NP_722577.2
XM_011532621.2 532 Silent Mutation GCC,GCT A8A XP_011530923.1
XM_011532622.2 532 Intron XP_011530924.1
XM_011532623.2 532 Intron XP_011530925.1
XM_017003505.1 532 Intron XP_016858994.1
XM_017003506.1 532 UTR 5 XP_016858995.1
Gene
EPT1
Gene Name
ethanolaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details