Product Details

SNP ID
rs201831015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86442185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCGAGTGGCCCTGGCTCTCCGG[A/G]ACCATTCGAGCTGTTTCCCACACCG
Phenotype
MIM: 611512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM3A PubMed Links

Gene Details

Gene
KDM3A
Gene Name
lysine demethylase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146688.1 196 Silent Mutation GGA,GGG G46G NP_001140160.1
NM_018433.5 196 Silent Mutation GGA,GGG G46G NP_060903.2
XM_006712051.3 196 UTR 5 XP_006712114.1
XM_017004493.1 196 Silent Mutation GGA,GGG G46G XP_016859982.1
XM_017004494.1 196 UTR 5 XP_016859983.1

View Full Product Details